Learn More
Quanterix Inc TSA PLUS FLUORESCIEN SYSTEM
SDP
Catalog No. 509049407
Encompass
TSA Plus Fluorescein System, for 50-150 Slides. These systems are designed for direct fluorescence detection in ISH or IHC procedures.
Catalog No. 50-904-9407 Supplier Quanterix Inc Supplier No. NEL741001KT
TSA Plus Fluorescein System, for 50-150 Slides. These systems are designed for direct fluorescence detection in ISH or IHC procedures.