Learn More
Quanterix Inc TSA Plus TMR 50-150 Slides
SDP
Catalog No. NC1816416
Encompass
TSA Plus tetramethylrhodamine (TMR) detection kit, for amplification of signal in immunohistochemistry (IHC), immunofluorescence (IF) or in situ hybridization protocols. This kit amplifies by depositing extra TMR in the localized area of the probe or antibody.
Catalog No. NC1816416 Supplier Quanterix Inc Supplier No. NEL742001KT
TSA Plus tetramethylrhodamine (TMR) detection kit, for amplification of signal in immunohistochemistry (IHC), immunofluorescence (IF) or in situ hybridization protocols. This kit amplifies by depositing extra TMR in the localized area of the probe or antibody.