Learn More
NEOGEN Tryptic Soy Agar (Soybean-Casein Digest Agar), 500 g
SDP
Catalog No. 502015159
Encompass_Preferred
Tryptic Soy Agar is used for the preparation and maintenance of test strains used in growth promotion tests, suitability of the counting methods and as negative controls as described in the Harmonized USP/EP/JP. It is also a support plating medium for various protocols described in the microbial enumeration test section of the Harmonized USP/EP/JP. Tryptic Soy Agar is not intended for use in the diagnosis of disease or other conditions in humans.Enzymatic digests of casein and soybean act as a source of nitrogen and amino acid and sodium chloride maintains the osmotic balance.Formula:Enzymatic Digest of Casein - 15.0 g/LEnzymatic Digest of Soybean - 5.0 g/LSodium Chloride - 5.0 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-agar
Catalog No. 50-201-5159 Supplier NEOGEN Supplier No. 700002950
Tryptic Soy Agar is used for the preparation and maintenance of test strains used in growth promotion tests, suitability of the counting methods and as negative controls as described in the Harmonized USP/EP/JP. It is also a support plating medium for various protocols described in the microbial enumeration test section of the Harmonized USP/EP/JP. Tryptic Soy Agar is not intended for use in the diagnosis of disease or other conditions in humans.Enzymatic digests of casein and soybean act as a source of nitrogen and amino acid and sodium chloride maintains the osmotic balance.Formula:Enzymatic Digest of Casein - 15.0 g/LEnzymatic Digest of Soybean - 5.0 g/LSodium Chloride - 5.0 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-agar