Learn More
MOLTOX Molecular Toxicology Inc Tryptic Soy Agar (TSA) w/Lecithin & Tween 80, pH 7.3 +/- 0.2, 100 mL/bottle, Store at Room Temperature
SDP
Catalog No. NC1005591
Encompass
Also known as TSA or Trypticase Soy Agar, Tryptic Soy Agar is used for the isolation and cultivation of fastidious and nonfastidious microorganisms. This medium can be used for a wide variety of purposes including maintenance of stock cultures, plate counts and microorganism isolation of a range of specimens. It is also used for testing bacterial contaminants in cosmetics. The addition of blood can be used for the visualization of hemolytic reactions produced by various bacterial species. The addition of Tween 80 (Polysorbate 80) and/or Lecithin, is used for environmental monitoring as they inactivate disinfectant residue. Whether you need a general-purpose, nonselective media or you need added blood to facilitate a more fasitdious bacterial lawn for the detection of hemolytic reactions, we have several products to choose from. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. NC1005591 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26739
Also known as TSA or Trypticase Soy Agar, Tryptic Soy Agar is used for the isolation and cultivation of fastidious and nonfastidious microorganisms. This medium can be used for a wide variety of purposes including maintenance of stock cultures, plate counts and microorganism isolation of a range of specimens. It is also used for testing bacterial contaminants in cosmetics. The addition of blood can be used for the visualization of hemolytic reactions produced by various bacterial species. The addition of Tween 80 (Polysorbate 80) and/or Lecithin, is used for environmental monitoring as they inactivate disinfectant residue. Whether you need a general-purpose, nonselective media or you need added blood to facilitate a more fasitdious bacterial lawn for the detection of hemolytic reactions, we have several products to choose from. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.