Learn More
MilliporeSigma TSA with LTHThio cont.-ICR+ | 200/pack
Catalog No. NC1763514
Encompass
Application: Environmental monitoring for isolators & clean rooms (surface and air monitoring), Detection of fungi and bacteria.
Catalog No. NC1763514 Supplier MilliporeSigma Supplier No. 1467830200
Application: Environmental monitoring for isolators & clean rooms (surface and air monitoring), Detection of fungi and bacteria.