Learn More
Biolegend TotalSeq™-A0452 anti-Nuclear Pore Complex Proteins Hashtag 2
Supplier:  Biolegend 682207
Encompass_Preferred
TotalSeq™-A0452 anti-Nuclear Pore Complex Proteins Hashtag 2 [MAb414]; Isotype: Mouse IgG1; Reactivity: Vertebrate, Xenopus, Yeast; Apps: PG; Size: 10 ug
Catalog No. 50-208-7444