Learn More
Biolegend TotalSeq™-A0459 anti-Nuclear Pore Complex Proteins Hashtag 9
Supplier:  Biolegend 682221
Encompass_Preferred
TotalSeq™-A0459 anti-Nuclear Pore Complex Proteins Hashtag 9 [MAb414]; Isotype: Mouse IgG1; Reactivity: Vertebrate, Xenopus, Yeast; Apps: PG; Size: 10 ug
Catalog No. 50-208-7451