Learn More
Surmodics Inc Tris Buffered Saline/Casein Block & Diluent -10X, 100mL
SDP
Catalog No. NC9388908
Encompass_Preferred
BioFX™ Tris Buffered Saline (TBS)/Casein Block and Diluent – 10X Concentrate minimizes non-specific interactions of the secondary reagents with each other and the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins. The solution is ready for dilution to specific application requirements.
Catalog No. NC9388908 Supplier Surmodics Inc Supplier No. TBSC010001
BioFX™ Tris Buffered Saline (TBS)/Casein Block and Diluent – 10X Concentrate minimizes non-specific interactions of the secondary reagents with each other and the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins. The solution is ready for dilution to specific application requirements.