Learn More
Description
- Borosilicate bottle
- Phenolic screw cap with septum
Specifications
Specifications
| Color | Amber |
| Media Type | Culture Media |
| Fill Volume | 100 mL |
| For Use With (Application) | Sterility testing |
| Packaging Type | Double bagged |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.