Learn More
NEOGEN Tryptic Soy Broth (Soybean-Casein Digest Broth), 10 kg
SDP
Catalog No. 502015167
Encompass_Preferred
Tryptic Soy Broth (Soybean-Casein Digest Broth) is used for the cultivation of a wide variety of microorganisms. Tryptic Soy Broth conforms to Harmonized USP/EP/JP requirements and is not intended for use in the diagnosis of disease or other conditions in humans.Tryptic Soy Broth, a general purpose medium, is commonly referred to as Casein Soya Bean or Soybean-Casein Digest broth, and abbreviated as TSB. This medium was originally developed for use without blood in determining the effectiveness of sulfonamides against pneumococci and other organisms. Clostridia and non-sporulating anaerobes grow luxuriantly in this broth when incubated under anaerobic conditions. TSB is recommended for testing bacterial contaminants in cosmetics and complies with established standards in the food industry.For more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-broth
Catalog No. 50-201-5167 Supplier NEOGEN Supplier No. 700002964
Tryptic Soy Broth (Soybean-Casein Digest Broth) is used for the cultivation of a wide variety of microorganisms. Tryptic Soy Broth conforms to Harmonized USP/EP/JP requirements and is not intended for use in the diagnosis of disease or other conditions in humans.Tryptic Soy Broth, a general purpose medium, is commonly referred to as Casein Soya Bean or Soybean-Casein Digest broth, and abbreviated as TSB. This medium was originally developed for use without blood in determining the effectiveness of sulfonamides against pneumococci and other organisms. Clostridia and non-sporulating anaerobes grow luxuriantly in this broth when incubated under anaerobic conditions. TSB is recommended for testing bacterial contaminants in cosmetics and complies with established standards in the food industry.For more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-broth