Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics TSB (Tryptic Soy Broth) with 15% Glycerol, 2mL
SDP

Catalog No. 23001734 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-734 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-734 Supplier Hardy Diagnostics Supplier No. D09
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tryptic Soy Broth

Specifications

Product Type Microbiology Media
Fill Volume 2 mL
No. per Pack 100
Packaging Type 2mL Cryovial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.