Learn More
ATCC TSB with 10% glycerol, Pkg of 12 (1.0ml/vial)
Catalog No. 502384164
Encompass
Trypticase soy broth (TSB) with 10% glycerol (v/v) is used as a freeze medium for storage of non-fastidious bacteria, fungi, and yeast. This product is provided in an individual unit of use (1.0 mL).
Catalog No. 50-238-4164 Supplier ATCC Supplier No. 202200
Trypticase soy broth (TSB) with 10% glycerol (v/v) is used as a freeze medium for storage of non-fastidious bacteria, fungi, and yeast. This product is provided in an individual unit of use (1.0 mL).