Learn More
MOLTOX Molecular Toxicology Inc Tryptic Soy Broth (TSB) w / 2% Tween 80 & 0.3% Lecithin, 90 mL/bottle, Store in Refrigerator
SDP
Catalog No. NC0484530
Encompass
This item has a minimum order quantity of 96 per supplier requirements. Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi. Whether you need a general-purpose, nonselective media or you need added blood to facilitate a more fasitdious bacterial lawn for the detection of hemolytic reactions, we have several products to choose from. MOLTOX does not provide microorganisms for testing. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. NC0484530 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26653
This item has a minimum order quantity of 96 per supplier requirements. Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi. Whether you need a general-purpose, nonselective media or you need added blood to facilitate a more fasitdious bacterial lawn for the detection of hemolytic reactions, we have several products to choose from. MOLTOX does not provide microorganisms for testing. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.