Learn More
Andwin Scientific TSC AGAR 500G
SDP
Catalog No. NC9920386
Encompass_Preferred
The EMD Millipore Dehydrated Culture MediaSimmons Citrate Agar is in powdered and granular form, which maintains homogeneity by capturing the medias original component distribution in each granule and preventing settling out. This product lessens the dangers linked with inhalation of fine dust and toxic substances. This product is sold in the quantity of 500 grams.
Catalog No. NC9920386 Supplier Andwin Scientific Supplier No. 1.11972.0500
The EMD Millipore Dehydrated Culture MediaSimmons Citrate Agar is in powdered and granular form, which maintains homogeneity by capturing the medias original component distribution in each granule and preventing settling out. This product lessens the dangers linked with inhalation of fine dust and toxic substances. This product is sold in the quantity of 500 grams.