Learn More
Aviva Systems Biology Corp TSC22D1 Antibody - middle region (ARP38726_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38726_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSC22D1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSC22D1. Alternate names: DKFZp686O19206, MGC17597, RP11-269C23.2, TGFB1I4, TSC22, Ptg-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56770