Learn More
Aviva Systems Biology Corp TSC22D2 Antibody - N-terminal region (ARP39137_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39137_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSC22D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TSC22D2. Alternate names: TILZ4a, TILZ4b, TILZ4cApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56917