Learn More
Aviva Systems Biology Corp TSC22D2 Antibody - C-terminal region (ARP39138_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39138_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSC22D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TSC22D2. Alternate names: TILZ4a, TILZ4b, TILZ4cApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56918