Learn More
Aviva Systems Biology Corp TSC22D2 Antibody - C-terminal region (ARP33698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSC22D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TSC22D2. Alternate names: KIAA0669, TILZ4a, TILZ4b, TILZ4cApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55119