Learn More
Aviva Systems Biology Corp Tsc22d4 Antibody - N-terminal region (ARP37339_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37339_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against Tsc22d4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of mouse Tsc22d4. Alternate names: 0610009M14Rik, 1700023B23Rik, Thg-1pit, Tilz2, AI415410ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56352