Learn More
Thermo Scientific™ TSE Series Under Counter Stacking Kit
Catalog No. 507548SK Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Undercounter Refrigerator/Freezer
Catalog No. For Use With (Equipment)
50-754-8SK Undercounter Refrigerator/Freezer
1 options
Catalog No. 50-754-8SK Supplier Thermo Scientific™ Supplier No. 507548SK
Add to Cart
Add to Cart
Stacking Kit Under Counter support bracket kit. Required for TSE Series Stackable Refrigerators and Freezers
TRUSTED_SUSTAINABILITY
Specifications
Type Support Bracket Kit
For Use With (Equipment) Undercounter Refrigerator/Freezer
Includes Front plate (1), Back plate (2), M5x15 Screws (3), T4x12 Screws (8)
Quantity 1 Kit