Learn More
Andwin Scientific TSE TEK CMPND OCT 4OZ BT,12/CS
SDP
Catalog No. NC9159334
Encompass_Preferred
The Tissue-Tek® optimum cutting temperature (O.C.T.) solution is composed of water-soluble glycols and resins that give a specimen matrix for cryostat sectioning at temperatures of –10°C (14°F) and below. This product does not leave any residue on slides during and after staining prcoess. It is packaged in a 118mL (4oz.) bottle and is sold in a case of 12.
This item is not returnable. View return policy
Catalog No. NC9159334 Supplier Andwin Scientific Supplier No. 4583X2
The Tissue-Tek® optimum cutting temperature (O.C.T.) solution is composed of water-soluble glycols and resins that give a specimen matrix for cryostat sectioning at temperatures of –10°C (14°F) and below. This product does not leave any residue on slides during and after staining prcoess. It is packaged in a 118mL (4oz.) bottle and is sold in a case of 12.