Learn More
New England Biolabs, Inc. TseI – 375 units
SDP
Catalog No. 50812303
Encompass
A restriction endonuclease that recognizes the sequence G^CWG_C.
Catalog No. 50-812-303 Supplier New England Biolabs, Inc. Supplier No. R0591L
A restriction endonuclease that recognizes the sequence G^CWG_C.