Learn More
Aviva Systems Biology Corp Tsen15 Antibody - C-terminal region (ARP52469_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52469_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tsen15. It was validated on Western Blot. . Alternate names: 5730449L18Rik, AL023077, Sen15ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-718