Learn More
ABclonal Technology TSEN54 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15227500UL
Encompass_Preferred
This gene encodes a subunit of the tRNA splicing endonuclease complex, which catalyzes the removal of introns from precursor tRNAs. The complex is also implicated in pre-mRNA 3-prime end processing. Mutations in this gene result in pontocerebellar hypoplasia type 2.
Catalog No. 50-001-04581