Learn More
Aviva Systems Biology Corp TSFM Antibody - N-terminal region (ARP32675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSFM. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TSFM. Alternate names: EFTS, EFTSMTApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54703