Learn More
Aviva Systems Biology Corp TSFM Antibody - middle region (ARP31455_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31455_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSFM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSFM. Alternate names: COXPD3, EF-TS, EF-Tsmt, EFTS, EFTSMTApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54230