Learn More
Genetex Inc Anti-TSG101 antibody [4A10]
SDP
Supplier:  Genetex Inc GTX70255
Encompass
The Anti-TSG101 antibody [4A10] from GeneTex is a Mouse Monoclonal antibody to TSG101. The Anti-TSG101 antibody [4A10] Recognizes Human, Mouse, Monkey, Rat, Hamster. The Anti-TSG101 antibody [4A10] has been validated for the following Applications: EM, ELISA, FACS, ICC/IF, IHC, IHC-P, IP, WB.
Catalog No. NC1221937