Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSG Series General Purpose Laboratory Refrigerators

Catalog No. TSG72RPGA
Change view
Click to view available options
Capacity (Metric):
764 L
1274 L
1387 L
2038 L
Door Style:
Glass
Sliding Glass
Cabinet Material:
Gray-painted
White-painted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Door Style Cabinet Material
TSG72RPGA 2038 L Glass White-painted
TSG30RPGA 764 L Glass White-painted
TSG45RPLA 1274 L Sliding Glass White-painted
TSG49RPGA 1387 L Glass White-painted
TSG49RSGA 1387 L Glass Gray-painted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSG72RPGA Supplier Thermo Scientific™ Supplier No. TSG72RPGA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Designed for your everyday laboratory needs

Thermo Scientific™ TSG Series general-purpose laboratory refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2° to 10°C controlled environment.

  • Featuring a wide range of cabinet sizes from 12 cu. ft. to 72 cu. ft.
  • Whisper-quiet operation
  • ENERGY STAR certifiedTemperature, door ajar*, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
  • Warranty: varies by country, please contact us for details
    *All models except TSG45RPLA and TSG45RSLA come equipped with door ajar alarm
TRUSTED_SUSTAINABILITY

Specifications

Description TSG Series general purpose laboratory refrigerator
Capacity (English) 72 cu. ft.
Capacity (Metric) 2038 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) +5°C
Refrigerant R290
Door Style Glass
No. of Doors 3
Shelves 12 Full
Capacity (English) Shelf 100 lb.
Capacity (Metric) Shelf 45 kg
Cabinet Material White-painted
Exterior Finish Painted
Interior Stainless Steel
Certifications/Compliance cULus, ENERGYSTAR, Prop 65
Control Type Digital Microprocessor
Ports 1 Rear access port
Warranty 2 years parts and labor, plus 3-year compressor warranty
Defrost Adaptive Defrost
Monitoring Options Alarm
Access Security Key Lock
Dimensions (D x W x H) Interior 28.5x72x61.75 in. (724x1829x1569 mm)
Dimensions (L x W x H) Exterior 32x75.7x81 in.(813x1924x2057 mm)
Amperage 9.9 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Green Features Energy efficient
Shipping Weight (English) 733 lb.
Shipping Weight (Metric) 332 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.