Learn More
Aviva Systems Biology Corp TSG101 Antibody - middle region (ARP37310_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37310_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TSG101. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: CC2, AI255943ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56347