Learn More
Aviva Systems Biology Corp TSG101 Antibody - C-terminal region (ARP38773_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38773_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TSG101. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TSG101. Alternate names: TSG10, VPS23ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56791