Learn More
Alpco Ltd Canine TSH ELISA
SDP
Supplier:  Alpco Ltd 55TSHCAE01
Encompass_Preferred
The Canine TSH ELISA is an enzyme immunoassay for the quantitative measurement of canine TSH (thyrotropin) in serum or EDTA plasma.
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3641