Learn More
Enzo Life Sciences TSH ELISA kit (1x96wells)
SDP
Catalog No. 502013826
Encompass_Preferred
The TSH ELISA kit is a colorimetric immunometric immunoassay kit with results in 75 minutes. Alternative name: Thyroid-stimulating hormone, Thyrotropin, hTSH. Assay Time: 75 min. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3826 Supplier Enzo Life Sciences Supplier No. ENZKIT1310001
The TSH ELISA kit is a colorimetric immunometric immunoassay kit with results in 75 minutes. Alternative name: Thyroid-stimulating hormone, Thyrotropin, hTSH. Assay Time: 75 min. Applications: ELISA, Colorimetric detection . Species reactivity: Human .