Learn More
Crystal Chem Inc Thyroid Stimulating Hormone (TSH) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80982
Encompass
The Thyroid Stimulating Hormone (TSH) ELISA is a highly sensitive assay that is used to quantify levels of human TSH in serum. Highlights: Kits use sample size of only 50 µL. High sensitivity of 0.1 µIU/mL. Dynamic range of 0.2 - 30 µIU/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7883