Learn More
Eurofins Discoverx TSH, Thyroid stimulating hormone, 10 µg. (92-1111)
Supplier:  Eurofins Discoverx 921111
Encompass
Thyroid stimulating hormone (TSH) is a glycoprotein thyroid regulating pituitary hormone that acts as an agonist of TSHR and stimulates metabolism in all tissues. Induces thyroid production of thyroxine and triiodothyronine. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9646