Learn More
Thermo Scientific™ TSHP Series FMS High-Performance Lab Refrigerators
Shop All Thermo Scientific Products

Description
Protect flammable materials, solvents and intermediates using Thermo Scientific™ TSHP FMS High-Performance Laboratory Refrigerators, which meet stringent U.S. Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.† These refrigerators are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. Each is listed by UL as a special purpose refrigerator intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†
Key Features
- Easy-to-use, intuitive digital display featuring microprocessor control
- Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
- Convenient access port and casters
- Outstanding construction featuring painted, high-quality steel exteriors with durable finish
- Ignition-free interior cabinet design for the safe storage of flammable materials
- Meets safety standards in accordance with UL, NFPA† and OSHA† for regulatory compliance
- Optional, built-in, inkless, seven-day graphic chart recorder
- Optional, barrier-protected 4-20 milliamp output for external monitoring
- Manual defrost
Ordering Information
Temperature chart recorders, and 4-20 milliamp are available separately. Not available outside North America.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Flammable Storage Refrigerator |
| Capacity (English) | 23 cu. ft. |
| Capacity (Metric) | 651 L |
| Temperature Range | 1°C to 8°C |
| Door Style | Solid |
| No. of Doors | 1 |
| Shelves | 4 |
| Cabinet Material | Spark-free Steel |
| Exterior Finish | Painted |
| Certifications/Compliance | UL |
| Show More |
† Products complying with the applicable requirements of NFPA 70, National Electric Code, NPFA 45, Fire Protection for Laboratories Using Chemicals, and NFPA 99 Health Care Facilities are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.