Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7253MSM5P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: TSHRB/1405. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a. Positive Control: 293T Cells. Thyroid gland. Cellular Staining: Cell Surface ;Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human TSHR, B-Chain protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 42kDa.Chromosome: 14q31. Gene Symbol: TSHR. Gene ID: 7253. Ensemble: ENSG00000165409. Uniprot: XP_005268094.
Catalog No. 50-188-5250