Learn More
Aviva Systems Biology Corp TSHR Antibody - N-terminal region (ARP41870_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41870_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TSHR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TSHR. Alternate names: LGR3, MGC75129, hTSHR-I, CHNG1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58027