Learn More
Carbosynth LLC. TSHR Blocking Peptide, 100 ug
SDP
Supplier:  Carbosynth LLC. 33R45020.1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
A synthetic peptide for use as a blocking control in assays to test for specificity of TSHR antibody, catalog no. 70R-2915
Catalog No. 50-187-0649