Learn More
Medchemexpress LLC TSI-01 | 704878-75-1 | 98.5% | 328.15 | 1 ML
SDP
Catalog No. 5000438497
Encompass_Preferred
TSI-01 is a selective inhibitor of LPCAT2, an enzyme involved in the rapid synthesis of pro-inflammatory phospholipid mediators like platelet-activating factor (PAF). It is designed for research use only.
  • Selectively inhibits lysophosphatidylcholine acyltransferase 2 (LPCAT2).
  • Demonstrates higher potency against human LPCAT2 than LPCAT1.
  • Suppresses PAF biosynthesis in mouse peritoneal macrophages.
  • Available in various package sizes for research flexibility.
  • Provided with detailed documentation for quality assurance.
  • Multiple solvent options for reconstitution are available.
Catalog No. 50-004-38497 Supplier Medchemexpress LLC Supplier No. HY11282510MM/1ML
TSI-01 is a selective inhibitor of LPCAT2, an enzyme involved in the rapid synthesis of pro-inflammatory phospholipid mediators like platelet-activating factor (PAF). It is designed for research use only.
  • Selectively inhibits lysophosphatidylcholine acyltransferase 2 (LPCAT2).
  • Demonstrates higher potency against human LPCAT2 than LPCAT1.
  • Suppresses PAF biosynthesis in mouse peritoneal macrophages.
  • Available in various package sizes for research flexibility.
  • Provided with detailed documentation for quality assurance.
  • Multiple solvent options for reconstitution are available.