Learn More
Tosoh Bioscience LLC TSKGEL ALPHA-5000 7.8X300
Supplier:  Tosoh Bioscience LLC 0018342
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel Alpha-5000; Media: Polymer; Particle: 10 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - Multi-Solvent Capability
Catalog No. 50-851-493