Learn More
Tosoh Bioscience LLC TSKGEL ALPHA GUARD COLUMN
Supplier:  Tosoh Bioscience LLC 0018345
Encompass
Column, Guard; For 7.8mm I.D. Tosoh TSKgel Alpha columns; Media: Polymer; 6mm I.D. x 4cm
Catalog No. 50-851-496