Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 1X150 5UM
Supplier: Tosoh Bioscience LLC 0021486
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 1.0mm x 15cm Length; Stainless Steel