Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 1X250 5UM
Supplier:  Tosoh Bioscience LLC 0021487
Encompass
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 1.0mm x 25cm Length; Stainless Steel
Catalog No. 50-851-691