Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80, 2MM ID X 150M
Supplier:  Tosoh Bioscience LLC 0021865
Encompass
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 3 micrometer; Pore Size: 80 angstrom; 2mm I.D. x 15cm Length; Stainless Steel
Catalog No. NC9674627