Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 4.6X100 5UM
Catalog No. 50851558 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 4.6mm I.D. x 10cm Length; Stainless Steel
Catalog No. 50-851-558 Supplier Tosoh Bioscience LLC Supplier No. 0019533
Column, Normal Phase/Hydrophilic Interaction Chromatography; Tosoh TSKgel Amide-80; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 4.6mm I.D. x 10cm Length; Stainless Steel