Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 GUARD 4X10
Catalog No. 50851534 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Guard; For 4.6mm I.D. Tosoh TSKgel Amide-80 columns; Media: Silica; 4.6mm I.D. x 1cm Length
This item is not returnable. View return policy
Catalog No. 50-851-534 Supplier Tosoh Bioscience LLC Supplier No. 0019021
Column, Guard; For 4.6mm I.D. Tosoh TSKgel Amide-80 columns; Media: Silica; 4.6mm I.D. x 1cm Length