Learn More
Tosoh Bioscience LLC TSKGEL AMIDE-80 GUARD COLUMN
Supplier:  Tosoh Bioscience LLC 0014461
Encompass
Column, Guard; For 21.5mm I.D. Tosoh TSKgel Amide-80 column; Media: Silica; 21.5mm I.D. x 7.5cm Length
Catalog No. 50-851-357