Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST 3SWXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0020026
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel BioAssist G3SWxl; Media: Silica; Particle: 5 micrometer; Pore Size: 250 angstrom; 7.8mm I.D. x 30cm Length; PEEK
Catalog No. 50-851-627