Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST 6PW 7.8X300
Supplier:  Tosoh Bioscience LLC 0020024
Encompass
Column, Gel Filtration Chromatography; Tosoh; TSKgel; BioAssist G6PW; Material: PEEK(housing), hydroxylated polymethacrylate(base); Media: polymer; Particle: 17u m; Pore Size: > 1000angstrom 7.8mm I.D. x 30cm
Catalog No. 50-851-625