Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST CHELATE
Supplier:  Tosoh Bioscience LLC 0020022
Encompass
Column, Affinity Chromatography; Tosoh; TSKgel; Matrix: BioAssist Chelate phase; Material: PEEK hardware; Media: polymer; Particle: 10u m; Applications: immunoglobulins, transferrin, lectins, membrane proteins; 7.8mm I.D. x 5cm length
Catalog No. 50-851-623